1-(2-Fluor-4-nitrophenyl)piperazin

Lieferant: FLUOROCHEM

010741-1G 010741-5G
FLUO010741-1GEA 148 EUR
FLUO010741-1G FLUO010741-5G
1-(2-Fluor-4-nitrophenyl)piperazin
1-(2-Fluor-4-nitrophenyl)piperazin
Formel: C₁₀H₁₂FN₃O₂
Molecular Weight: 225.22 g/mol
MDL: MFCD01572463
CAS-Nummer: 154590-33-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR