N-(4-Bromphenyl)maleimid

Lieferant: FLUOROCHEM

011109-25G 011109-100G
FLUO011109-25GEA 0 EUR
FLUO011109-25G FLUO011109-100G
N-(4-Bromphenyl)maleimid
N-(4-Bromphenyl)maleimid
Formel: C₁₀H₆BrNO₂
Schmelzpunkt: 128,5…129.5 °C
MDL: MFCD00030664
CAS-Nummer: 13380-67-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR