(2,3-Xylyloxy)essigsäure

Lieferant: FLUOROCHEM

Synonyme: (2,3-Dimethylphenoxy)essigsäure

011162-25G 011162-100G 011162-5G
FLUO011162-25GEA 0 EUR
FLUO011162-25G FLUO011162-100G FLUO011162-5G
(2,3-Xylyloxy)essigsäure
(2,3-Xylyloxy)essigsäure
Formel: (C₆H₃(CH₃)₂)OCH₂COOH
Molecular Weight: 180,2 g/mol
MDL: MFCD00016814
CAS-Nummer: 2935-63-9
EINECS: 220-911-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR