3-Indolylessigsäurehydrazid

Lieferant: FLUOROCHEM

Synonyme: Indol-3-ylessigsäurehydrazid, Indol-3-essigsäurehydrazid

011172-5G 011172-25G
FLUO011172-5GEA 79.1 EUR
FLUO011172-5G FLUO011172-25G
3-Indolylessigsäurehydrazid
3-Indolylessigsäurehydrazid
Formel: C₁₀H₁₁N₃O
Molecular Weight: 189.22 g/mol
Schmelzpunkt: 198
MDL: MFCD00005634
CAS-Nummer: 5448-47-5
EINECS: 226-672-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR