2-Amino-4,6-dichlorpyrimidin

Lieferant: FLUOROCHEM

Synonyme: 4,6-Dichloropyrimidin-2-ylamine

011355-100G 011355-25G
FLUO011355-100GEA 76.2 EUR
FLUO011355-100G FLUO011355-25G
2-Amino-4,6-dichlorpyrimidin
2-Amino-4,6-dichloropyrimidine
Formel: C₄H₃Cl₂N₃
Molecular Weight: 163,99 g/mol
Schmelzpunkt: 216…220 °C
MDL: MFCD00006090
CAS-Nummer: 56-05-3
EINECS: 200-253-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR