2-Amino-5-methylthiazol
Lieferant: FLUOROCHEM
Synonyme:
5-Methylthiazol-2-ylamin
011366-100G
011366-25G
FLUO011366-100GEA
121
EUR
FLUO011366-100G
FLUO011366-25G
2-Amino-5-methylthiazol
2-Amino-5-methylthiazole
Formel:
C₄H₆N₂S Molecular Weight: 114,17 g/mol Siedepunkt: 222 °C (1013 hPa) Schmelzpunkt: 96 °C |
MDL:
MFCD00078317 CAS-Nummer: 7305-71-7 EINECS: 423-800-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...