3,6-Difluorphthalsäureanhydrid

Lieferant: CHEMPUR

Synonyme: 3,6-Difluor-1,2-benzoldicarbonsäureanhydrid

FL13001.POR 013001-1G 013001-5G
CHMPFL13001.POREA 0 EUR
CHMPFL13001.POR FLUO013001-1G FLUO013001-5G
3,6-Difluorphthalsäureanhydrid
3,6-Difluorphthalsäureanhydrid
Formel: C₈H₂F₂O₃
MDL: MFCD00134537
CAS-Nummer: 652-40-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR