6-Fluor-o-anisaldehyd

Lieferant: FLUOROCHEM

Synonyme: 6-Fluor-2-anisaldehyd, 2-Fluor-6-methoxybenzaldehyd

013017-1G 013017-5G
FLUO013017-1GEA 31.1 EUR
FLUO013017-1G FLUO013017-5G
6-Fluor-o-anisaldehyd
6-Fluor-o-anisaldehyd
Formel: C₈H₇FO₂
Molecular Weight: 154.14 g/mol
MDL: MFCD01090998
CAS-Nummer: 146137-74-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR