3,5-Dimethylbenzolsulfonylchlorid

Lieferant: CHEMPUR

Synonyme: m-Xylol-5-sulfonsäurechlorid, m-Xylol-5-sulfonylchlorid, 3,5-Dimethylbenzol-1-sulfonylchlorid, 3,5-Dimethylbenzolsulfonsäurechlorid, 3,5-Dimethylbenzolsulfochlorid

FL13066.POR 013066-1G 013066-25G 013066-5G
CHMPFL13066.POREA 0 EUR
CHMPFL13066.POR FLUO013066-1G FLUO013066-25G FLUO013066-5G
3,5-Dimethylbenzolsulfonylchlorid
3,5-Dimethylbenzolsulfonylchlorid
Formel: C₈H₉ClO₂S
Molecular Weight: 204.68 g/mol
MDL: MFCD03094655
CAS-Nummer: 2905-27-3
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR