4-Brom-2-chlorbenzolsulfonylchlorid
Lieferant: FLUOROCHEM
Synonyme:
4-Brom-2-chlorbenzolsulfonsäurechlorid, 4-Brom-2-chlorbenzolsulfochlorid
013081-5G
013081-25G
FLUO013081-5GEA
82.1
EUR
FLUO013081-5G
FLUO013081-25G
4-Brom-2-chlorbenzolsulfonylchlorid
4-Brom-2-chlorbenzolsulfonylchlorid
Formel:
C₆H₃BrCl₂O₂S Molecular Weight: 289,96 g/mol Schmelzpunkt: 76…81 °C |
MDL:
MFCD03094630 CAS-Nummer: 351003-52-6 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...