2-Amino-5-phenylpyridin

Lieferant: CHEMPUR

Synonyme: 5-Phenyl-2-pyridylamin, 5-Phenylpyridin-2-amin

FL13115.POR 013115-5G 013115-1G
CHMPFL13115.POREA 0 EUR
CHMPFL13115.POR FLUO013115-5G FLUO013115-1G
2-Amino-5-phenylpyridin
2-Amino-5-phenylpyridine
Formel: C₁₁H₁₀N₂
Molecular Weight: 170,21 g/mol
MDL: MFCD01692452
CAS-Nummer: 33421-40-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR