Benzoylacetonitril

Lieferant: FLUOROCHEM

Synonyme: 3-Phenyl-3-ketopropionsäurenitril, 3-Phenyl-3-oxopropionsäurenitril

013117-2G 013117-10G
FLUO013117-2GEA 0 EUR
FLUO013117-2G FLUO013117-10G
Benzoylacetonitril
Benzoylacetonitril
Formel: (C₆H₅)C(O)CH₂CN
Molecular Weight: 145.16
Siedepunkt: 301
Schmelzpunkt: 83,5
MDL: MFCD00001942
CAS-Nummer: 614-16-4
EINECS: 210-369-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR