6-Chlornicotinonitril

Lieferant: FLUOROCHEM

Synonyme: 2-Chlor-5-pyridincarbonitril, 6-Chlornicotinsäurenitril, 2-Chlor-5-cyanpyridin, 2-Chlor-5-pyridincarbonsäurenitril

CFF390.5 CFF390.10 013153-5G 013153-25G 013153-1G
CHMPCFF390.5EA 799 EUR
CHMPCFF390.5 CHMPCFF390.10 FLUO013153-5G FLUO013153-25G FLUO013153-1G
6-Chlornicotinonitril
6-Chlornicotinonitril
Formel: C₆H₃ClN₂
Molecular Weight: 138,56 g/mol
MDL: MFCD00084941
CAS-Nummer: 33252-28-7
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR