2-Brom-4-fluorbenzaldehyd

Lieferant: CHEMPUR

FL13205.POR 013205-25G
CHMPFL13205.POREA 0 EUR
CHMPFL13205.POR FLUO013205-25G
2-Brom-4-fluorbenzaldehyd
2-Bromo-4-fluorobenzaldehyde
Formel: C₇H₄BrFO
Molecular Weight: 203.01 g/mol
MDL: MFCD00672923
CAS-Nummer: 59142-68-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR