2-Brom-5-fluorbenzaldehyd

Lieferant: CHEMPUR

FL13206.POR 013206-25G 013206-5G 013206-100G
CHMPFL13206.POREA 0 EUR
CHMPFL13206.POR FLUO013206-25G FLUO013206-5G FLUO013206-100G
2-Brom-5-fluorbenzaldehyd
2-Bromo-5-fluorobenzaldehyde
Formel: C₇H₄BrFO
MDL: MFCD00142872
CAS-Nummer: 94569-84-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR