2-Brom-4-fluor-1-iodbenzol

Lieferant: CHEMPUR
Achtung

Synonyme: 1-Brom-5-fluor-2-iodbenzol, 3-Brom-1-fluor-4-iodbenzol

FL13234.POR 013234-25G
CHMPFL13234.POREA 0 EUR
CHMPFL13234.POR FLUO013234-25G
2-Brom-4-fluor-1-iodbenzol
2-Brom-4-fluor-1-iodbenzol
Formel: C₆H₃BrFI
Molecular Weight: 300,9 g/mol
MDL: MFCD00142868
CAS-Nummer: 202865-73-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR