6-Brom-o-toluidin

Lieferant: CHEMPUR

FL13450.POR 013450-1G 013450-5G
CHMPFL13450.POREA 0 EUR
CHMPFL13450.POR FLUO013450-1G FLUO013450-5G
6-Brom-o-toluidin
6-Brom-o-toluidin
Formel: C₇H₈BrN
Molecular Weight: 186,05 g/mol
MDL: MFCD04039885
CAS-Nummer: 53848-17-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR