2,5-Bis(trifluormethyl)benzonitril

Lieferant: CHEMPUR

FL13760.POR 013760-25G 013760-5G
CHMPFL13760.POREA 0 EUR
CHMPFL13760.POR FLUO013760-25G FLUO013760-5G
2,5-Bis(trifluormethyl)benzonitril
2,5-Bis(trifluormethyl)benzonitril
Formel: C₉H₃F₆N
Molecular Weight: 239.12 g/mol
MDL: MFCD03094423
CAS-Nummer: 51012-27-2
UN: 3276
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR