2,5-Bis(trifluormethyl)benzylbromid

Lieferant: CHEMPUR

Synonyme: 1-(Brommethyl)-2,5-bis(trifluormethyl)benzol

FL13764.POR 013764-25G 013764-5G
CHMPFL13764.POREA 0 EUR
CHMPFL13764.POR FLUO013764-25G FLUO013764-5G
2,5-Bis(trifluormethyl)benzylbromid
2,5-Bis(trifluormethyl)benzylbromid
Formel: C₉H₅BrF₆
Molecular Weight: 307,03 g/mol
MDL: MFCD00799485
CAS-Nummer: 302911-98-4
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR