2-Brom-4-(trifluormethyl)pyridin

Lieferant: FLUOROCHEM

013908-5G 013908-1G
FLUO013908-5GEA 137 EUR
FLUO013908-5G FLUO013908-1G
2-Brom-4-(trifluormethyl)pyridin
2-Bromo-4-(trifluoromethyl)pyridine
Formel: C₆H₃BrF₃N
Molecular Weight: 226 g/mol
MDL: MFCD00153085
CAS-Nummer: 175205-81-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR