3-Iod-4-methoxybenzoesäure

Lieferant: FLUOROCHEM

Synonyme: 3-Iodo-p-anisic acid

015259-1G 015259-25G 015259-5G
FLUO015259-1GEA 31.1 EUR
FLUO015259-1G FLUO015259-25G FLUO015259-5G
3-Iod-4-methoxybenzoesäure
3-Iod-4-methoxybenzoesäure
Formel: C₈H₇IO₃
Molecular Weight: 278,05 g/mol
MDL: MFCD00017347
CAS-Nummer: 68507-19-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR