2-Brom-5-nitrothiazol
Lieferant: FLUOROCHEM
017215-5G
017215-10G
FLUO017215-5GEA
128
EUR
FLUO017215-5G
FLUO017215-10G
2-Brom-5-nitrothiazol
2-Bromo-5-nitrothiazole
Formel:
C₃HBrN₂O₂S Molecular Weight: 209.02 g/mol Schmelzpunkt: 93 Dichte: 2.295 g/cm³ (20 °C) |
MDL:
MFCD00005317 CAS-Nummer: 3034-48-8 EINECS: 221-226-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...