5-Methyl-3-isoxazolcarbonsäure

Lieferant: FLUOROCHEM

CFF179.10 CFF179.25 017252-25G 017252-1G 017252-5G
CHMPCFF179.10EA 532 EUR
CHMPCFF179.10 CHMPCFF179.25 FLUO017252-25G FLUO017252-1G FLUO017252-5G
5-Methyl-3-isoxazolcarbonsäure
5-Methylisoxazole-3-carboxylic acid
Formel: C₅H₅NO₃
Molecular Weight: 127.1 g/mol
MDL: MFCD01318162
CAS-Nummer: 3405-77-4
EINECS: 222-289-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR