5-Isoxazolcarbonsäure

Lieferant: FLUOROCHEM

Synonyme: Isoxazole-5-carboxylic acid

CFF303.2.5 CFF303.10 017345-2.5G 017345-10G
CHMPCFF303.2.5EA 268 EUR
CHMPCFF303.2.5 CHMPCFF303.10 FLUO017345-2.5G FLUO017345-10G
5-Isoxazolcarbonsäure
5-Isoxazolecarboxylic acid
Formel: C₄H₃NO₃
Molecular Weight: 113,07 g/mol
MDL: MFCD00156151
CAS-Nummer: 21169-71-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR