1-(2-Chlor-6-fluorbenzyl)piperazin

Lieferant: FLUOROCHEM

CFF311.5 017351-5G 017351-1G
CHMPCFF311.5EA 532 EUR
CHMPCFF311.5 FLUO017351-5G FLUO017351-1G
1-(2-Chlor-6-fluorbenzyl)piperazin
1-(2-Chlor-6-fluorbenzyl)piperazin
Formel: C₁₁H₁₄ClFN₂
Molecular Weight: 228,7 g/mol
MDL: MFCD00816605
CAS-Nummer: 215655-20-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR