1-(2-Fluorbenzyl)piperazin

Lieferant: FLUOROCHEM

CFF368.5 CFF368.10 CFF368.25 017397-5G 017397.10G 017397-25G 017397-1G
CHMPCFF368.5EA 532 EUR
CHMPCFF368.5 CHMPCFF368.10 CHMPCFF368.25 FLUO017397-5G FLUO017397.10G FLUO017397-25G FLUO017397-1G
1-(2-Fluorbenzyl)piperazin
1-(2-Fluorbenzyl)piperazin
Formel: C₁₁H₁₅FN₂
Molecular Weight: 194,25 g/mol
MDL: MFCD02256027
CAS-Nummer: 89292-78-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR