3-Amino-2-methoxypyridin
Lieferant: FLUOROCHEM
Synonyme:
2-Methoxy-3-pyridylamin, 2-Methoxypyridin-3-amin
CFF378.5
CFF378.10
CFF378.25
017404-5G
017404-10G
017404-25G
017404-1G
CHMPCFF378.5EA
401
EUR
CHMPCFF378.5
CHMPCFF378.10
CHMPCFF378.25
FLUO017404-5G
FLUO017404-10G
FLUO017404-25G
FLUO017404-1G
3-Amino-2-methoxypyridin
3-Amino-2-methoxypyridine
Formel:
C₆H₈N₂O Molecular Weight: 124,14 g/mol |
MDL:
MFCD00833386 CAS-Nummer: 20265-38-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...