3-Chlor-2-thenoesäure

Lieferant: FLUOROCHEM

Synonyme: 3-Chlorthiophen-2-carbonsäure, 3-Chlor-2-thiophencarbonsäure

CFF381.5 CFF381.10 CFF381.25 017407-5G 017407-10G 017407-25G
CHMPCFF381.5EA 532 EUR
CHMPCFF381.5 CHMPCFF381.10 CHMPCFF381.25 FLUO017407-5G FLUO017407-10G FLUO017407-25G
3-Chlor-2-thenoesäure
3-Chlor-2-thenoesäure
Formel: C₅H₃ClO₂S
Molecular Weight: 162,6 g/mol
Schmelzpunkt: 187…188 °C
MDL: MFCD00043888
CAS-Nummer: 59337-89-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR