3-(Methylthio)anilin
Lieferant: FLUOROCHEM
Synonyme:
3-(Methylmercapto)anilin
018141-25G
018141-100G
FLUO018141-25GEA
84.9
EUR
FLUO018141-25G
FLUO018141-100G
3-(Methylthio)anilin
3-(Methylthio)anilin
Formel:
C₇H₉NS Siedepunkt: 293 °C (1013 hPa) Dichte: 1,1464 g/cm³ (20 °C) |
MDL:
MFCD00007793 CAS-Nummer: 1783-81-9 EINECS: 217-232-5 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...