2-Amino-5-chlorpyrimidin

Lieferant: FLUOROCHEM

Synonyme: 5-Chlorpyrimidin-2-ylamin

018177-1G 018177-5G
FLUO018177-1GEA 31.1 EUR
FLUO018177-1G FLUO018177-5G
2-Amino-5-chlorpyrimidin
2-Amino-5-chloropyrimidine
Formel: C₄H₄ClN₃
Molecular Weight: 129.55 g/mol
MDL: MFCD00014604
CAS-Nummer: 5428-89-7
EINECS: 226-581-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR