3-Benzylanilin

Lieferant: FLUOROCHEM

018206-1G 018206-5G
FLUO018206-1GEA 130 EUR
FLUO018206-1G FLUO018206-5G
3-Benzylanilin
3-Benzylanilin
Formel: C₁₃H₁₃N
Molecular Weight: 183,25 g/mol
MDL: MFCD01310782
CAS-Nummer: 61424-26-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR