2-Brom-4-chloranisol

Lieferant: FLUOROCHEM

Synonyme: 2-Bromo-4-chloro-1-methoxybenzene

018250-100G 018250-25G
FLUO018250-100GEA 112 EUR
FLUO018250-100G FLUO018250-25G
2-Brom-4-chloranisol
2-Brom-4-chloranisol
Formel: C₇H₆BrClO
Molecular Weight: 221,48 g/mol
MDL: MFCD00079705
CAS-Nummer: 60633-25-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR