1-Brom-3-chlor-4-iodbenzol
Lieferant: FLUOROCHEM
Synonyme:
4-Brom-2-chlor-1-iodbenzol
018257-5G
018257-100G
018257-25G
FLUO018257-5GEA
0
EUR
FLUO018257-5G
FLUO018257-100G
FLUO018257-25G
1-Brom-3-chlor-4-iodbenzol
4-Bromo-2-chloro-1-iodobenzene
Formel:
C₆H₃BrClI Molecular Weight: 317,35 g/mol |
MDL:
MFCD00079709 CAS-Nummer: 31928-47-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...