1-(2-Bromphenyl)-2-thioharnstoff
Lieferant: FLUOROCHEM
|
Gefahr
|
Synonyme:
N-(2-Bromphenyl)thioharnstoff, 2-Bromphenylthioharnstoff
018317-1G
018317-5G
FLUO018317-1GEA
58.1
EUR
FLUO018317-1G
FLUO018317-5G
1-(2-Bromphenyl)-2-thioharnstoff
1-(2-Bromphenyl)-2-thioharnstoff
Formel:
(C₆H₄Br)NHC(S)NH₂ Molecular Weight: 231.12 g/mol Siedepunkt: 319 Schmelzpunkt: 130…132.5 °C |
MDL:
MFCD00041146 CAS-Nummer: 5391-30-0 UN: 2811 ADR: 6.1 I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...