3-(o-Tolyl)propionsäure
Lieferant: FLUOROCHEM
Synonyme:
3-(2-Methylphenyl)propansäure, 3-(2-Methylphenyl)propionsäure
018855-1G
018855-5G
FLUO018855-1GEA
69.8
EUR
FLUO018855-1G
FLUO018855-5G
3-(o-Tolyl)propionsäure
3-(o-Tolyl)propionsäure
Formel:
C₁₀H₁₂O₂ |
MDL:
MFCD00079773 CAS-Nummer: 22084-89-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...