3-(m-Tolyl)propionsäure

Lieferant: FLUOROCHEM

Synonyme: 3-(3-Methylphenyl)propionsäure, 3-(3-Methylphenyl)propansäure

018856-1G 018856-5G
FLUO018856-1GEA 67.1 EUR
FLUO018856-1G FLUO018856-5G
3-(m-Tolyl)propionsäure
3-(m-Tolyl)propionsäure
Formel: C₁₀H₁₂O₂
Molecular Weight: 164.2 g/mol
MDL: MFCD00060326
CAS-Nummer: 3751-48-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR