1-(2-Nitrophenyl)-2-thioharnstoff
Lieferant: FLUOROCHEM
|
Gefahr
|
Synonyme:
N-(2-Nitrophenyl)thiourea, 2-Nitrophenylthiourea
018920-1G
018920-5G
FLUO018920-1GEA
69.8
EUR
FLUO018920-1G
FLUO018920-5G
1-(2-Nitrophenyl)-2-thioharnstoff
1-(2-Nitrophenyl)-2-thioharnstoff
Formel:
(C₆H₄(NO₂))NC(S)NH₂ Molecular Weight: 197.22 g/mol Schmelzpunkt: 140.5…141.5 °C |
MDL:
MFCD00041237 CAS-Nummer: 51039-84-0 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...