3-Fluor-N-methylanilin

Lieferant: FLUOROCHEM

019287-10G 019287-25G
FLUO019287-10GEA 0 EUR
FLUO019287-10G FLUO019287-25G
3-Fluor-N-methylanilin
3-Fluor-N-methylanilin
Formel: C₇H₈FN
Molecular Weight: 125.15 g/mol
MDL: MFCD02683095
CAS-Nummer: 1978-37-6
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR