5-(2-Bromphenyl)-1H-tetrazol

Lieferant: CHEMPUR

Synonyme: 5-(2-Bromophenyl)tetrazole

FL19493.POR 019493-5G 019493-1G
CHMPFL19493.POREA 0 EUR
CHMPFL19493.POR FLUO019493-5G FLUO019493-1G
5-(2-Bromphenyl)-1H-tetrazol
5-(2-Bromphenyl)-1H-tetrazol
Formel: C₇H₅BrN₄
Molecular Weight: 225.05 g/mol
Schmelzpunkt: 183…184 °C
MDL: MFCD00151799
CAS-Nummer: 73096-42-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR