1-Methyl-3-phenylpiperazin
Lieferant: FLUOROCHEM
|
Gefahr
|
Synonyme:
MEFPIP
021556-5G
021556-1G
FLUO021556-5GEA
48.8
EUR
FLUO021556-5G
FLUO021556-1G
1-Methyl-3-phenylpiperazin
1-Methyl-3-phenylpiperazin
Formel:
C₁₁H₁₆N₂ Molecular Weight: 176,26 g/mol |
MDL:
MFCD03411603 CAS-Nummer: 5271-27-2 EINECS: 431-180-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...