4-Chlor-3-fluorbenzylamin
Lieferant: FLUOROCHEM
Synonyme:
(4-Chloro-3-fluorophenyl)methanamine
021807-5G
021807-1G
FLUO021807-5GEA
254
EUR
FLUO021807-5G
FLUO021807-1G
4-Chlor-3-fluorbenzylamin
4-Chlor-3-fluorbenzylamin
Formel:
C₇H₇ClFN Molecular Weight: 159.59 g/mol |
MDL:
MFCD04116008 CAS-Nummer: 72235-58-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...