2,3-Dimethoxyanilin
Lieferant: FLUOROCHEM
Synonyme:
(2,3-Dimethoxyphenyl)amine
021813-1G
021813-5G
FLUO021813-1GEA
72.9
EUR
FLUO021813-1G
FLUO021813-5G
2,3-Dimethoxyanilin
2,3-Dimethoxyanilin
Formel:
C₈H₁₁NO₂ Molecular Weight: 153,18 g/mol |
MDL:
MFCD00180769 CAS-Nummer: 6299-67-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...