3,5-Dimethylisoxazol

Lieferant: FLUOROCHEM

023218-250G 023218-25G 023218-100G
FLUO023218-250GEA 321 EUR
FLUO023218-250G FLUO023218-25G FLUO023218-100G
3,5-Dimethylisoxazol
3,5-Dimethylisoxazole
Formel: C₅H₇NO
Molecular Weight: 97,12 g/mol
Siedepunkt: 142 °C (1013 hPa)
Dichte: 0,98 g/cm³ (20 °C)
MDL: MFCD00003156
CAS-Nummer: 300-87-8
EINECS: 206-100-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR