3,5-Dimethyl-4-iodisoxazol

Lieferant: FLUOROCHEM

023250-5G 023250-25G
FLUO023250-5GEA 0 EUR
FLUO023250-5G FLUO023250-25G
3,5-Dimethyl-4-iodisoxazol
3,5-Dimethyl-4-iodoisoxazole
Formel: C₅H₆INO
Molecular Weight: 223,01 g/mol
MDL: MFCD00173743
CAS-Nummer: 10557-85-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR