2-Amino-4-methylthiazol
Lieferant: FLUOROCHEM
Synonyme:
4-Methylthiazol-2-ylamin
023295-100G
023295-25G
FLUO023295-100GEA
152
EUR
FLUO023295-100G
FLUO023295-25G
2-Amino-4-methylthiazol
2-Amino-4-methylthiazole
Formel:
C₄H₆N₂S Molecular Weight: 114,17 g/mol Siedepunkt: 231 °C (1013 hPa) Schmelzpunkt: 42 °C Dichte: 1,202 g/cm³ (50 °C) |
MDL:
MFCD00005329 CAS-Nummer: 1603-91-4 EINECS: 216-505-6 Merck Index: 13,00449 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...