Uracil, weißes Pulver
Lieferant: MP Biomedicals
Synonyme:
2,4-Pyrimidindion, 2,4-Dihydroxypyrimidin, Pyrimidin-2,4(1H,3H)-dion
0210320491
0210320425
0210320480
ICNA0210320491EA
790
EUR
ICNA0210320491
ICNA0210320425
ICNA0210320480
Uracil, weißes Pulver
Uracil
Uracil is a pyrimidine derivative.
Used in drug delivery and in pharmaceuticals. In addition to this, it is used in the synthesis of caffeine and in the determination of microbial contamination of tomatoes.
Soluble in NaOH (40 mg/ml - clear, colourless solution, in hot water, ammonia water and other alkalies; slightly soluble in cold water; almost insoluble in alcohol and ether.
Formel:
C₄H₄N₂O₂ Molecular Weight: 112,09 g/mol Siedepunkt: 379 °C (1013 hPa) Schmelzpunkt: >300 °C (Lit.) Dichte: 0,99744 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006016 CAS-Nummer: 66-22-8 EINECS: 200-621-9 Merck Index: 12,09985 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...