1,8-Cineol ~99%, klare, farblose Flüssigkeit
Lieferant: MP Biomedicals
|
Achtung
|
Synonyme:
1,3,3-Trimethyl-2-oxabicyclo[2.2.2]octan, 1,8-Cineol, Limonen-1,8-oxid, 1,8-Epoxy-p-menthan
0215068990
0215068950
ICNA0215068990EA
0
EUR
ICNA0215068990
ICNA0215068950
1,8-Cineol ~99%, klare, farblose Flüssigkeit
Eucalyptol
Eucalyptol (cineole) is an ingredient in many brands of mouthwash and cough suppressant. It is used as an insecticide and insect repellent.It is used in flavorings, fragrances, and cosmetics.
Soluble in alcohol, most fixed oils, organic solvents, glycerin, propylene glycol, chloroform and ether; insoluble in water.
Formel:
C₁₀H₁₈O Molecular Weight: 154,25 g/mol Siedepunkt: 174…177 °C Schmelzpunkt: 1,5 °C Dichte: 0,921…0.924 g/cm³ (20 °C) Flammpunkt: 50 °C (Lit.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00167977 CAS-Nummer: 470-82-6 EINECS: 207-431-5 UN: 1993 ADR: 3,III Merck Index: 12,03940 |
Spezifikation Testergebnisse
Identity Test | Passes |
Purity | ~99% |
FTIR | Conforms to Standard |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...