1-(3,5-Dichlorphenyl)-2-thioharnstoff
Lieferant: MENAI
|
Gefahr
|
Synonyme:
N-(3,5-Dichlorphenyl)thioharnstoff, 3,5-Dichlorphenylthioharnstoff
DH587.50
DH587.100
DH587.5
MENADH587.50EA
0
EUR
MENADH587.50
MENADH587.100
MENADH587.5
1-(3,5-Dichlorphenyl)-2-thioharnstoff
1-(3,5-Dichlorphenyl)-2-thioharnstoff
Formel:
(C₆H₃Cl₂)NC(S)NH₂ Molecular Weight: 221.11 g/mol Siedepunkt: 326 Schmelzpunkt: 180 |
MDL:
MFCD00041168 CAS-Nummer: 107707-33-5 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...