1-(4-Fluorphenyl)-2-thioharnstoff
Lieferant: MENAI
|
Gefahr
|
Synonyme:
4-Fluorphenylthioharnstoff, N-(4-Fluorphenyl)thioharnstoff
DH588.25
DH588.100
DH588.5
MENADH588.25EA
0
EUR
MENADH588.25
MENADH588.100
MENADH588.5
1-(4-Fluorphenyl)-2-thioharnstoff
1-(4-Fluorphenyl)-2-thioharnstoff
Formel:
(C₆H₄F)NHC(S)NH₂ Molecular Weight: 170.21 g/mol Siedepunkt: 276 Schmelzpunkt: 169…170 °C |
MDL:
MFCD00041180 CAS-Nummer: 459-05-2 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...