1-(3,4-Dichlorphenyl)-2-thioharnstoff
Lieferant: MENAI
|
Gefahr
|
Synonyme:
N-(3,4-Dichlorphenyl)thioharnstoff, 3,4-Dichlorphenylthioharnstoff
DH613.50
DH613.10
MENADH613.50EA
0
EUR
MENADH613.50
MENADH613.10
1-(3,4-Dichlorphenyl)-2-thioharnstoff
1-(3,4-Dichlorphenyl)-2-thioharnstoff
Formel:
(C₆H₃Cl₂)NC(S)NH₂ Molecular Weight: 221.11 g/mol Siedepunkt: 326 Schmelzpunkt: 220 |
MDL:
MFCD00004935 CAS-Nummer: 19250-09-0 EINECS: 242-919-1 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...